Få mere at vide
7-Hydroxy Aminopterin, TRC
Artikelnummer. 30407621
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30407621 10 mg
30404780 1 mg
30404160 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30407621 Leverandør Toronto Research Chemicals Leverandørnr. TRCH79725010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97772-99-1
Molekylær formel C19H20N8O6
Molekylvægt (g/mol) 456.41
Synonym N-[4-[[(2,4-Diamino-7,8-dihydro-7-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic Acid, N-[p-[[(2,4-Diamino-7-hydroxy-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic Acid, N-[4-[[(2,4-Diamino-1,7-dihydro-7-oxo-6-pteridinyl)methyl]amino]benzoyl]- L-Glutamic Acid
IUPAC navn (2S)-2-[[4-[(2,4-diamino-7-oxo-8H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMIL C1=CC(=CC=C1C(=O)N[C@@H](CCC(=O)O)C(=O)O)NCC2=NC3=C(NC2=O)N=C(N=C3N)N
Tekniske data
Kemisk navn eller materiale 7-Hydroxy Aminopterin
Mængde 10 mg
InChI formel InChI=1S/C19H20N8O6/c20-14-13-15(27-19(21)25-14)26-17(31)11(23-13)7-22-9-3-1-8(2-4-9)16(30)24-10(18(32)33)5-6-12(28)29/h1-4,10,22H,5-7H2,(H,24,30)(H,28,29)(H,32,33)(H5,20,21,25,26,27,31)/t10-/m0/s1
Formel vægt 456.1506