Få mere at vide
7-Hydroxy-3-oxo-Cholest-4-en-26-oic Acid, TRC
Artikelnummer. 30138622
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30138622 25 mg -
30005455 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138622 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94966025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 115538-85-7
Molekylær formel C27H42O4
Molekylvægt (g/mol) 430.62
Synonym 7alpha-Hydroxy-3-oxo-4-cholestenoic Acid, 7alpha-Hydroxy-3-oxocholest-4-en-26-oic Acid
IUPAC navn (6R)-6-[(7R,8S,9S,10R,13R,14S,17R)-7-hydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]-2-methylheptanoic acid
SMIL C[C@H](CCCC(C)C(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 7-Hydroxy-3-oxo-Cholest-4-en-26-oic Acid
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C27H42O4/c1-16(6-5-7-17(2)25(30)31)20-8-9-21-24-22(11-13-27(20,21)4)26(3)12-10-19(28)14-18(26)15-23(24)29/h14,16-17,20-24,29H,5-13,15H2,1-4H3,(H,30,31)/t16-,17?,20-,21+,22+,23-,24+,26+,27-/m1/s1
Formel vægt 430.3083
Procent renhed >95%