Få mere at vide
7-Hydroxy-1,3,6-naphthalenetrisulfonic Acid Trisodium, TRC
Artikelnummer. 30005909
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005909 25 mg
30003270 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005909 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95072525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 53683-45-7
Molekylær formel C10 H5 O10 S3 . 3 Na
Molekylvægt (g/mol) 450.3
Synonym 2-Hydroxy-3,6,8-naphthalenetrisulfonic Acid Trisodium Salt, 2-Naphthol-3,6,8-trisulfonic Acid Trisodium
IUPAC navn trisodium;7-hydroxynaphthalene-1,3,6-trisulfonate
SMIL [Na+].[Na+].[Na+].Oc1cc2c(cc(cc2cc1S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-]
Tekniske data
Kemisk navn eller materiale 7-Hydroxy-1,3,6-naphthalenetrisulfonic Acid Trisodium
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C10H8O10S3.3Na/c11-8-4-7-5(2-10(8)23(18,19)20)1-6(21(12,13)14)3-9(7)22(15,16)17;;;/h1-4,11H,(H,12,13,14)(H,15,16,17)(H,18,19,20);;;/q;3*+1/p-3
Formel vægt 449.8738
Procent renhed >95%