Få mere at vide
7-Ethyl-d3-camptothecin, TRC
Artikelnummer. 30370880
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30370880 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370880 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9008321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Ethyl-d3-camptothecin
CAS 1217626-02-2
Molekylær formel C22H17D3N2O4
Mængde 1 mg
Synonym (4S)-4,11-Diethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione-d3, 7-Ethyl-d3-20(S)-camptothecin, SN 22-d3
InChI formel InChI=1S/C22H20N2O4/c1-3-12-13-7-5-6-8-17(13)23-19-14(12)10-24-18(19)9-16-15(20(24)25)11-28-21(26)22(16,27)4-2/h5-9,27H,3-4,10-11H2,1-2H3/t22-/m0/s1/i1D3
SMIL O=C1C2=C([C@@](O)(CC)C(OC2)=O)C=C3C4=NC5=CC=CC=C5C(CC([2H])([2H])[2H])=C4CN31
IUPAC navn (S)-4-ethyl-11-(ethyl-2,2,2-d3)-4-hydroxy-1,12-dihydro-14H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H)-dione
Molekylvægt (g/mol) 379.42
Formel vægt 379.16