missing translation for 'onlineSavingsMsg'
Få mere at vide

7-Ethyl-10-(4-N-aminopentanoic acid)-1-piperidino)carbonyloxycamptothecin, TRC

Artikelnummer. 30385272
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30385272 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30385272 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8989005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 7-Ethyl-10-(4-N-aminopentanoic acid)-1-piperidino)carbonyloxycamptothecin
CAS 181467-56-1
Molekylær formel C33 H38 N4 O8
Mængde 5 mg
InChI formel InChI=1S/C33H38N4O8/c1-3-21-22-15-20(45-32(42)36-13-10-19(11-14-36)34-12-6-5-7-28(38)39)8-9-26(22)35-29-23(21)17-37-27(29)16-25-24(30(37)40)18-44-31(41)33(25,43)4-2/h8-9,15-16,19,34,43H,3-7,10-14,17-18H2,1-2H3,(H,38,39)/t33-/m0/s1
SMIL CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(OC(=O)N6CCC(CC6)NCCCCC(=O)O)cc15)[C@@](O)(CC)C(=O)OC4
Molekylvægt (g/mol) 618.6768
Formel vægt 618.269
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.