Få mere at vide
7-Ethyl-10-(4-N-aminopentanoic acid)-1-piperidino)carbonyloxycamptothecin-d3, TRC
Artikelnummer. 30379085
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30379085 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379085 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8989021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Ethyl-10-(4-N-aminopentanoic acid)-1-piperidinocarbonyloxycamptothecin-d3
CAS 1217816-76-6
Molekylær formel C33 H35 D3 N4 O8
Mængde 1 mg
Synonym 4-[(4-Carboxybutyl)amino]-1-piperidinecarboxylic Acid (4S)-4,11-Diethyl -3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl Ester-d3, APC-d3, RPR 121056-d3
InChI formel InChI=1S/C33H38N4O8/c1-3-21-22-15-20(45-32(42)36-13-10-19(11-14-36)34-12-6-5-7-28(38)39)8-9-26(22)35-29-23(21)17-37-27(29)16-25-24(30(37)40)18-44-31(41)33(25,43)4-2/h8-9,15-16,19,34,43H,3-7,10-14,17-18H2,1-2H3,(H,38,39)/t33-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])CC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)[C@@]5(CC)O)OC(=O)N6CCC(CC6)NCCCCC(=O)O
Molekylvægt (g/mol) 621.7
Formel vægt 621.2878