Få mere at vide
7-Ethoxycoumarin-d5, TRC
Artikelnummer. 30374841
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30374841 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374841 Leverandør Toronto Research Chemicals Leverandørnr. TRCE891727100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Ethoxycoumarin-d5
CAS 1189956-39-5
Molekylær formel C11 2H5 H5 O3
Mængde 100 mg
Synonym 7-(1,1,2,2,2-Pentadeuterioethoxy)chromen-2-one, 7-(Ethoxy-1,1,2,2,2-d5)-2H-1-benzopyran-2-one, 7-Ethoxycoumarin-d5, 7-Ethoxy-d5-coumarin, 7-Ethoxycoumarin-D5 (ethoxy-D5), Ethyl-d5-umbelliferone
InChI formel InChI=1S/C11H10O3/c1-2-13-9-5-3-8-4-6-11(12)14-10(8)7-9/h3-7H,2H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1ccc2C=CC(=O)Oc2c1
IUPAC navn 7-(1,1,2,2,2-pentadeuterioethoxy)chromen-2-one
Molekylvægt (g/mol) 195.23
Formel vægt 195.0944