Få mere at vide
7-Desmethyl-3-hydroxyagomelatine, TRC
Artikelnummer. 30351133
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30351133 2.5 mg
30351953 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30351133 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2821502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 166527-00-0
Molekylær formel C14H15NO3
Molekylvægt (g/mol) 245.27
Synonym N-[2-(3,7-dihydroxy-1-naphthalenyl)ethyl]acetamide, Agomelatine 7-Desmethyl-3-Hydroxy Impurity
IUPAC navn N-(2-(3,7-dihydroxynaphthalen-1-yl)ethyl)acetamide
SMIL OC1=CC2=C(CCNC(C)=O)C=C(O)C=C2C=C1
Tekniske data
Kemisk navn eller materiale 7-Desmethyl-3-hydroxyagomelatine
Mængde 2.5 mg
InChI formel InChI=1S/C14H15NO3/c1-9(16)15-5-4-11-7-13(18)6-10-2-3-12(17)8-14(10)11/h2-3,6-8,17-18H,4-5H2,1H3,(H,15,16)
Formel vægt 245.11