Få mere at vide
7-Dechloro Griseofulvin, TRC
Artikelnummer. 30368778
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368778 25 mg
30367506 10 mg
30357166 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368778 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22645025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3680-32-8
Molekylær formel C17 H18 O6
Molekylvægt (g/mol) 318.32
Synonym (1'S,6'R)-2',4,6-Trimethoxy-6'-methyl-spiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, Spiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, 2',4,6-trimethoxy-6'-methyl- (6CI,7CI,8CI), Spiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, 2',4,6-trimethoxy-6'-methyl-, (1'S-trans)-, 7-Dechlorogriseofulvin, Dechlorogriseofulvin
IUPAC navn (2S,5'R)-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione
SMIL COC1=CC(=O)C[C@@H](C)[C@]12Oc3cc(OC)cc(OC)c3C2=O
Tekniske data
Kemisk navn eller materiale Dechlorogriseofulvin
Mængde 25 mg
InChI formel InChI=1S/C17H18O6/c1-9-5-10(18)6-14(22-4)17(9)16(19)15-12(21-3)7-11(20-2)8-13(15)23-17/h6-9H,5H2,1-4H3/t9-,17+/m1/s1
Formel vægt 318.1103