Få mere at vide
7-Cyanomethotrexate Dimethyl Ester-D3, TRC
Artikelnummer. 30352789
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30352789 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352789 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9818222.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Cyanomethotrexate Dimethyl Ester-D3
CAS 432545-61-4
Molekylær formel C23H22D3N9O5
Mængde 2.5 mg
Synonym N-[4-[[(2,4-Diamino-7-cyano-6-pteridinyl)methyl]methyl-d3-amino]benzoyl]-L-glutamic Acid Dimethyl Ester
InChI formel InChI=1S/C23H25N9O5/c1-32(11-16-15(10-24)28-20-18(27-16)19(25)30-23(26)31-20)13-6-4-12(5-7-13)21(34)29-14(22(35)37-3)8-9-17(33)36-2/h4-7,14H,8-9,11H2,1-3H3,(H,29,34)(H4,25,26,28,30,31)/t14-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])N(Cc1nc2c(N)nc(N)nc2nc1C#N)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)OC)C(=O)OC
IUPAC navn dimethyl (2S)-2-[[4-[(2,4-diamino-7-cyanopteridin-6-yl)methyl-(trideuteriomethyl)amino]benzoyl]amino]pentanedioate
Molekylvægt (g/mol) 510.52
Formel vægt 510.2167