Learn More
7-Coumaryl Triflate, TRC
Product Code. 30355246
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30355246 100 mg 100mg
1 options
Product Code. 30355246 Supplier Toronto Research Chemicals Supplier No. TRCC756450100MG
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-Coumaryl Triflate
CAS 108530-10-5
Molecular Formula C10H5F3O5S
Quantity 100 mg
Synonym 1,1,1-Trifluoro-methanesulfonic Acid 2-Oxo-2H-1-benzopyran-7-yl Ester
InChI Formula InChI=1S/C10H5F3O5S/c11-10(12,13)19(15,16)18-7-3-1-6-2-4-9(14)17-8(6)5-7/h1-5H
SMILES FC(F)(F)S(=O)(=O)Oc1ccc2C=CC(=O)Oc2c1
IUPAC Name (2-oxochromen-7-yl) trifluoromethanesulfonate
Molecular Weight (g/mol) 294.2
Formula Weight 293.981