Få mere at vide
7-Chloromethyl Drospirenone, TRC
Artikelnummer. 30136693
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136693 5 mg
30134355 100 mg
17987994 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136693 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3669805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 932388-90-4
Molekylær formel C24 H31 Cl O3
Molekylvægt (g/mol) 402.95
Synonym (7β,15α,16α,17α)-7-(Chloromethyl)-15,16-dihydro-17-hydroxy-3-oxo-3'H-cyclopropa[15,16]pregna-4,15-diene-21-carboxylic acid γ-lactone, 3'-Chloro-3',6-secodrospirenone, Drospirenone Imp. G (EP), 3'H-Cyclopropa[15,16]pregna-4,15-diene-21-carboxylic acid, 7-(chloromethyl)-15,16-dihydro-17-hydroxy-3-oxo-, γ-lactone, (7β,15α,16α,17α)-
IUPAC navn (1'R,2'S,3'S,5S,5'S,7'S,10'S,11'R,18'S)-18'-(chloromethyl)-7',11'-dimethylspiro[oxolane-5,6'-pentacyclo[8.8.0.02,7.03,5.011,16]octadec-15-ene]-2,14'-dione
SMIL C[C@]12CCC(=O)C=C1C[C@H](CCl)[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3[C@@H]5C[C@@H]5[C@@]46CCC(=O)O6
Tekniske data
Kemisk navn eller materiale 7-Chloromethyl Drospirenone
Mængde 5 mg
InChI formel InChI=1S/C24H31ClO3/c1-22-6-3-15(26)10-14(22)9-13(12-25)20-17(22)4-7-23(2)21(20)16-11-18(16)24(23)8-5-19(27)28-24/h10,13,16-18,20-21H,3-9,11-12H2,1-2H3/t13-,16-,17+,18+,20-,21+,22+,23+,24+/m1/s1
Formel vægt 402.1962