Få mere at vide
7-Chloro-5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide (Demoxepam), Mikromol™
Beskrivelse
7-Chloro-5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide (Demoxepam)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Demoxepam |
| CAS | 963-39-3 |
| Analyt- eller komponentnavne | Demoxepam |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H11 Cl N2 O2 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Chlordiazepoxide Hydrochloride Imp. A (EP), Chlordiazepoxide Imp. A (EP), Oxazepam Imp. E (EP), Chlordiazepoxide USP Related Compound A, Chlordiazepoxide USP RC A, 7-Chloro-5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide, Demoxepam, Chlordiazepoxide Related Compound A, Chlordiazepoxide Hydrochloride Impurity A, Chlordiazepoxide Impurity A, Oxazepam Impurity E, Chlordiazepoxide Impurity A |
| InChI formel | InChI=1S/C15H11ClN2O2/c16-11-6-7-13-12(8-11)15(10-4-2-1-3-5-10)18(20)9-14(19)17-13/h1-8H,9H2,(H,17,19) |
| SMIL | [O-][N+]1=C(c2ccccc2)c3cc(Cl)ccc3NC(=O)C1 |
| IUPAC navn | 7-chloro-4-oxido-5-phenyl-1,3-dihydro-1,4-benzodiazepin-4-ium-2-one |
| Vis mere |