Få mere at vide
7-Chloro-5-(2-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Mikromol™
Beskrivelse
7-Chloro-5-(2-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Desalkylflurazepam |
| CAS | 2886-65-9 |
| Analyt- eller komponentnavne | N-Desalkylflurazepam |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H10 Cl F N2 O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 7-Chloro-5-(2-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Midazolam Imp. F (EP), 1-Des[(diethylamino)ethyl]flurazepam, Flurazepam Monohydrochloride Imp. B (EP), Desalkylflurazepam, Flurazepam Imp. B (EP), Flurazepam USP Related Compound F, Flurazepam USP RC F, 2H-1,4-Benzodiazepin-2-one, 7-chloro-5-(2-fluorophenyl)-1,3-dihydro-, 2H-1,4-Benzodiazepin-2-one, 7-chloro-5-(o-fluorophenyl)-1,3-dihydro- (7CI,8CI), 7-Chloro-1,3-dihydro-5-(2'-fluorophenyl)-2H-1,4-benzodiazapin-2-one, 7-Chloro-5-(2'-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, 7-Chloro-5-(o-fluorophenyl)-1,3-dihydro-1,4-benzodiazepin-2-one, 7-Chloro-5-(o-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepine-2-one, CM 7116, Dealkylflurazepam, Descarbethoxyloflazepate, N-1-Desalkylflurazepam, N-Desalkyl-2-oxoquazepam, N-Desalkylflutoprazepam, Norfludiazepam, Norflurazepam, Norflutoprazepam, Ro 5-3367, Sch 17514, Flurazepam Related Compound F, Flurazepam Monohydrochloride Impurity B, Midazolam Impurity F, Flurazepam Impurity B |
| InChI formel | InChI=1S/C15H10ClFN2O/c16-9-5-6-13-11(7-9)15(18-8-14(20)19-13)10-3-1-2-4-12(10)17/h1-7H,8H2,(H,19,20) |
| SMIL | Fc1ccccc1C2=NCC(=O)Nc3ccc(Cl)cc23 |
| IUPAC navn | 7-chloro-5-(2-fluorophenyl)-1,3-dihydro-1,4-benzodiazepin-2-one |
| Vis mere |