Få mere at vide
7-Chloro-5-(2-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Mikromol™
Artikelnummer. 17903946
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17903946 25 mg
17942256 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17903946 Leverandør Mikromol Leverandørnr. MM0070.020025
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-Chloro-5-(2-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desalkylflurazepam
CAS 2886-65-9
Analyt- eller komponentnavne N-Desalkylflurazepam
Forsendelsestilstand Room Temperature
Molekylær formel C15 H10 Cl F N2 O
Certificeringer/compliance ISO 17025
Synonym 7-Chloro-5-(2-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Midazolam Imp. F (EP), 1-Des[(diethylamino)ethyl]flurazepam, Flurazepam Monohydrochloride Imp. B (EP), Desalkylflurazepam, Flurazepam Imp. B (EP), Flurazepam USP Related Compound F, Flurazepam USP RC F, 2H-1,4-Benzodiazepin-2-one, 7-chloro-5-(2-fluorophenyl)-1,3-dihydro-, 2H-1,4-Benzodiazepin-2-one, 7-chloro-5-(o-fluorophenyl)-1,3-dihydro- (7CI,8CI), 7-Chloro-1,3-dihydro-5-(2'-fluorophenyl)-2H-1,4-benzodiazapin-2-one, 7-Chloro-5-(2'-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, 7-Chloro-5-(o-fluorophenyl)-1,3-dihydro-1,4-benzodiazepin-2-one, 7-Chloro-5-(o-fluorophenyl)-1,3-dihydro-2H-1,4-benzodiazepine-2-one, CM 7116, Dealkylflurazepam, Descarbethoxyloflazepate, N-1-Desalkylflurazepam, N-Desalkyl-2-oxoquazepam, N-Desalkylflutoprazepam, Norfludiazepam, Norflurazepam, Norflutoprazepam, Ro 5-3367, Sch 17514, Flurazepam Related Compound F, Flurazepam Monohydrochloride Impurity B, Midazolam Impurity F, Flurazepam Impurity B
InChI formel InChI=1S/C15H10ClFN2O/c16-9-5-6-13-11(7-9)15(18-8-14(20)19-13)10-3-1-2-4-12(10)17/h1-7H,8H2,(H,19,20)
SMIL Fc1ccccc1C2=NCC(=O)Nc3ccc(Cl)cc23
IUPAC navn 7-chloro-5-(2-fluorophenyl)-1,3-dihydro-1,4-benzodiazepin-2-one
Molekylvægt (g/mol) 288.7
Mængde 25 mg
Formel vægt 288.0466 g/mol