Learn More
7-Chloro-4-hydroxyquinoline-3-carboxylic Acid, TRC
Product Code. 30352652
Change view
Click to view available options
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30352652 500 mg
30359917 100 mg
30360221 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30352652 Supplier Toronto Research Chemicals Supplier No. TRCC382335500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 86-47-5
Molekylær formel C10H6ClNO3
Molekylvægt (g/mol) 223.61
IUPAC navn 7-chloro-4-oxo-1H-quinoline-3-carboxylic acid
SMIL OC(=O)C1=CNc2cc(Cl)ccc2C1=O
Specifications
Kemisk navn eller materiale 7-Chloro-4-hydroxyquinoline-3-carboxylic Acid
Mængde 500 mg
InChI formel InChI=1S/C10H6ClNO3/c11-5-1-2-6-8(3-5)12-4-7(9(6)13)10(14)15/h1-4H,(H,12,13)(H,14,15)
Formel vægt 223.0036