Få mere at vide
7-Chloro-1-ethyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid, 90%, TRC
Artikelnummer. 30134491
Skift visning
Mængde:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134491 10 g
30133423 1 g
30010880 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134491 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36604510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68077-26-9
Molekylær formel C12 H9 Cl F N O3
Molekylvægt (g/mol) 269.66
Synonym 7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Norfloxacin Imp. A (EP)
IUPAC navn 7-chloro-1-ethyl-6-fluoro-4-oxoquinoline-3-carboxylic acid
SMIL CCN1C=C(C(=O)O)C(=O)c2cc(F)c(Cl)cc12
Tekniske data
Kemisk navn eller materiale 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Mængde 10 g
InChI formel InChI=1S/C12H9ClFNO3/c1-2-15-5-7(12(17)18)11(16)6-3-9(14)8(13)4-10(6)15/h3-5H,2H2,1H3,(H,17,18)
Formel vægt 269.0255