Få mere at vide
7-Chloro-1-Ethyl-6-Fluoro-1,4-Dihydro-4-Oxo-3-Quinolinecarboxylic Acid Ethyl Ester, TRC
Artikelnummer. 30358137
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358137 10 mg
30350810 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30358137 Leverandør Toronto Research Chemicals Leverandørnr. TRCC31551010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70458-94-5
Molekylær formel C14 H13 Cl F N O3
Molekylvægt (g/mol) 297.71
Synonym 7-Chloro-1-ethyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester, Ethyl 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, Pefloxacin Mesilate Dihydrate Imp. G (EP)
IUPAC navn ethyl 7-chloro-1-ethyl-6-fluoro-4-oxoquinoline-3-carboxylate
SMIL CCOC(=O)C1=CN(CC)c2cc(Cl)c(F)cc2C1=O
Tekniske data
Kemisk navn eller materiale Ethyl 7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Mængde 10 mg
InChI formel InChI=1S/C14H13ClFNO3/c1-3-17-7-9(14(19)20-4-2)13(18)8-5-11(16)10(15)6-12(8)17/h5-7H,3-4H2,1-2H3
Formel vægt 297.0568