Få mere at vide
7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid, Mikromol™
Artikelnummer. 30051046
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30051046 50 mg
1 options
Artikelnummer. 30051046 Leverandør Mikromol Leverandørnr. MM0018.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid
CAS 133210-96-5
Analyt- eller komponentnavne 7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C17 H18 Cl N3 O3
Certificeringer/compliance ISO 9001
Synonym Ciprofloxacin Hydrochloride Imp. D (EP), Ciprofloxacin Imp. D (EP), Ciprofloxacin USP Related Compound A (AS), Ciprofloxacin USP RC A (AS), 7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid, Ciprofloxacin Related Compound A, Ciprofloxacin Hydrochloride Impurity D, Ciprofloxacin Impurity D
InChI formel InChI=1S/C17H18ClN3O3/c18-13-8-14-11(7-15(13)20-5-3-19-4-6-20)16(22)12(17(23)24)9-21(14)10-1-2-10/h7-10,19H,1-6H2,(H,23,24)
SMIL OC(=O)C1=CN(C2CC2)c3cc(Cl)c(cc3C1=O)N4CCNCC4
IUPAC navn 7-chloro-1-cyclopropyl-4-oxo-6-piperazin-1-ylquinoline-3-carboxylic acid
Molekylvægt (g/mol) 347.8
Mængde 50 mg
Formel vægt 347.1037 g/mol