missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals

Catalog Number 15490519
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
82771-60-6
C9H10ClN
167.64
MFCD05861543
BLRKQLICTKRDDZ-UHFFFAOYSA-N
7-chloro-1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 7-chloro-1,2,3,4-tetrahydro, 7-chhloro-1,2,3,4-tetrahydroisoquinoline, 7-chloranyl-1,2,3,4-tetrahydroisoquinoline, isoquinoline,7-chloro-1,2,3,4-tetrahydro
12595069
7-chloro-1,2,3,4-tetrahydroisoquinoline
ClC1=CC2=C(CCNC2)C=C1
This item is not returnable. View return policy
82771-60-6
C9H10ClN
167.64
MFCD05861543
BLRKQLICTKRDDZ-UHFFFAOYSA-N
7-chloro-1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 7-chloro-1,2,3,4-tetrahydro, 7-chhloro-1,2,3,4-tetrahydroisoquinoline, 7-chloranyl-1,2,3,4-tetrahydroisoquinoline, isoquinoline,7-chloro-1,2,3,4-tetrahydro
12595069
7-chloro-1,2,3,4-tetrahydroisoquinoline
ClC1=CC2=C(CCNC2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 167.64
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydroisoquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'