Learn More
7-Bromo-4-methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 154264-95-6 |
| Molecular Formula | C9H10BrNO |
| MDL Number | MFCD02681913 |
| Quantity | 1 g |
| SMILES | CN1C2=CC=C(Br)C=C2OCC1 |
| Molecular Weight (g/mol) | 228.09 |
| Percent Purity | 95% |
| Chemical Name or Material | 7-Bromo-4-methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine |