Få mere at vide
7-Benzyloxy-N-des[[2-(2-hydroxy)ethoxy]ethyl] Quetiapine-d8, TRC
Artikelnummer. 30332738
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30332738 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30332738 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28593710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Benzyloxy-N-des[(2-(2-hydroxy)ethoxy)ethyl] Quetiapine-d8
CAS 1185177-12-1
Molekylær formel C24H15D8N3OS
Mængde 10 mg
SMIL [2H]C(NC([2H])([2H])C1([2H])[2H])([2H])C([2H])([2H])N1C2=NC3=CC=C(OCC4=CC=CC=C4)C=C3SC5=C2C=CC=C5
Molekylvægt (g/mol) 409.57