Learn More
7-Aminoheptanoic acid, 98%, Thermo Scientific Chemicals
Catalog Number p-7029994
Click to view available options
Quantity:
250 mg
1 g
5 g
929-17-9
C7H15NO2
145.202
MFCD00008242
XDOLZJYETYVRKV-UHFFFAOYSA-N
7-aminoenanthic acid, heptanoic acid, 7-amino, omega-aminoenantic acid, aminoenanthic acid, amino-enanthylic acid, 7-amino-heptanoic acid, 7-aminohepentanoic acid, omega-aminoheptanoic acid, unii-b10hjx2t48, kyselina 7-aminoenanthova czech
13580
7-aminoheptanoic acid
C(CCCN)CCC(=O)O
929-17-9
C7H15NO2
145.202
MFCD00008242
XDOLZJYETYVRKV-UHFFFAOYSA-N
7-aminoenanthic acid, heptanoic acid, 7-amino, omega-aminoenantic acid, aminoenanthic acid, amino-enanthylic acid, 7-amino-heptanoic acid, 7-aminohepentanoic acid, omega-aminoheptanoic acid, unii-b10hjx2t48, kyselina 7-aminoenanthova czech
13580
7-aminoheptanoic acid
C(CCCN)CCC(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 194°C to 196°C
Quantity 250 mg
Beilstein 906887
Formula Weight 145.19
Percent Purity 98%
Chemical Name or Material 7-Aminoheptanoic acid