missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Aminodesacetoxycephalosporanic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
7-Aminodesacetoxycephalosporanic acid is the Cephalexin metabolite. It is a Potent inhibitor of bacterial cell-wall enzyme. It is used in the synthesis of cephalosporins and for bioconversion studies.
Solubility
Soluble in water (partly), 1M ammonium hydroxide (50 mg/ml), and DMSO (Sparingly).
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
7-Aminodesacetoxycephalosporanic acid is the Cephalexin metabolite. It is a Potent inhibitor of bacterial cell-wall enzyme. It is used in the synthesis of cephalosporins and for bioconversion studies.
Solubility
Soluble in water (partly), 1M ammonium hydroxide (50 mg/ml), and DMSO (Sparingly).
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | ∽234°C (decomposition) |
Quantity | 250 mg |
Beilstein | 5273616 |
Solubility Information | Soluble in water (partly),1M ammonium hydroxide (50mg/ml),and DMSO (Sparingly). |
Formula Weight | 214.24 |
Percent Purity | 98% |
Chemical Name or Material | 7-Aminodesacetoxycephalosporanic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'