missing translation for 'onlineSavingsMsg'
Få mere at vide

7-Aminobenzo[a]pyrene, TRC

Artikelnummer. 30312133
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312133 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30312133 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5928505MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 7-Aminobenzo[a]pyrene
CAS 72297-05-3
Molekylær formel C20 H13 N
Mængde 5 mg
Synonym Benzo[a]pyren-7-amine, 7-Aminobenzo[a]pyrene
InChI formel InChI=1S/C20H13N/c21-18-6-2-5-15-16-10-9-13-4-1-3-12-7-8-14(11-17(15)18)20(16)19(12)13/h1-11H,21H2
SMIL Nc1cccc2c1cc3ccc4cccc5ccc2c3c45
IUPAC navn benzo[a]pyren-7-amine
Molekylvægt (g/mol) 267.32
Formel vægt 267.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.