missing translation for 'onlineSavingsMsg'
Learn More

7-(Acetyloxy)-2-chloro-dibenz[b,f][1,4]oxazepin-11(10H)-one, TRC

Product Code. 30326188
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30326188 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326188 Supplier Toronto Research Chemicals Supplier No. TRCA18743310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 7-(Acetyloxy)-2-chloro-dibenz[b,f][1,4]oxazepin-11(10H)-one
CAS 51370-03-7
Molekylær formel C15H10ClNO4
Mængde 10 mg
Synonym 2-Chloro-11-oxo-10,11-dihydrodibenzo[b,f][1,4]oxazepin-7-yl Acetate
InChI formel InChI=1S/C15H10ClNO4/c1-8(18)20-10-3-4-12-14(7-10)21-13-5-2-9(16)6-11(13)15(19)17-12/h2-7H,1H3,(H,17,19)
SMIL CC(=O)Oc1ccc2NC(=O)c3cc(Cl)ccc3Oc2c1
IUPAC navn (8-chloro-6-oxo-5H-benzo[b][1,4]benzoxazepin-2-yl) acetate
Molekylvægt (g/mol) 303.7
Formel vægt 303.0298
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.