Få mere at vide
7,8-Dihydro-1-naphthalenol, TRC
Artikelnummer. 30010403
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010403 250 mg
30009551 1 g
17903135 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010403 Leverandør Toronto Research Chemicals Leverandørnr. TRCD451130250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51927-48-1
Molekylær formel C10 H10 O
Molekylvægt (g/mol) 146.19
Synonym 1-Naphthalenol, 7,8-dihydro-, 1-Naphthol, 7,8-dihydro- (6CI), 7,8-Dihydro-1-naphthalenol, 7,8-Dihydro-1-naphthol, 8-Hydroxy-1,2-dihydronaphthalene, 7,8-Dihydronaphthalen-1-ol
IUPAC navn 7,8-dihydronaphthalen-1-ol
SMIL Oc1cccc2C=CCCc12
Tekniske data
Kemisk navn eller materiale 7,8-Dihydro-1-naphthalenol
Mængde 250 mg
InChI formel InChI=1S/C10H10O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1,3-5,7,11H,2,6H2
Formel vægt 146.0732