Få mere at vide
7,8-Diaminopelargonic Acid Dihydrochloride, TRC
Artikelnummer. 30138442
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30138442 5 mg -
30132482 2.5 mg 2.5mg
30008110 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138442 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4165505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 951786-35-9
Molekylær formel C9 H20 N2 O2 . 2 H Cl
Molekylvægt (g/mol) 261.19
Synonym (7R,8S)-7,8-Diaminononanoic Acid Hydrochloride
IUPAC navn (7R,8S)-7,8-diaminononanoic acid;dihydrochloride
SMIL Cl.Cl.C[C@H](N)[C@H](N)CCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale 7,8-Diaminopelargonic acid dihydrochloride
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C9H20N2O2.2ClH/c1-7(10)8(11)5-3-2-4-6-9(12)13;;/h7-8H,2-6,10-11H2,1H3,(H,12,13);2*1H/t7-,8+;;/m0../s1
Formel vægt 260.1058
Procent renhed >95%