Få mere at vide
7,8,9,10-Dehydro Doxorubicinone (~70%), TRC
Artikelnummer. 30362974
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362974 5 mg
30352489 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30362974 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2295805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1159977-23-7
Molekylær formel C21H14O8
Molekylvægt (g/mol) 394.33
Synonym 7,8,9,10-Dehydro Doxorubicinone
SMIL COC1=CC=CC2=C1C(=C3C(=C2O)C(=O)C4=CC(=CC(=C4C3=O)O)C(=O)CO)O
Tekniske data
Kemisk navn eller materiale 7,8,9,10-Dehydro Doxorubicinone
Mængde 5 mg
InChI formel InChI=1S/C21H14O8/c1-29-13-4-2-3-9-15(13)21(28)17-16(18(9)25)19(26)10-5-8(12(24)7-22)6-11(23)14(10)20(17)27/h2-6,22-23,25,28H,7H2,1H3