Få mere at vide
7,7',8,8'-Tetradehydro-4,5alpha:4',5'alpha-diepoxy-17,17'-dimethyl-2,2'-bimorphinanyl-3,3',6alpha,6'alpha-tetrol (Pseudomorphine; 2,2'-Bismorphine), Mikromol™
Artikelnummer. 17953076
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17953076 100 mg
17944106 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17953076 Leverandør Mikromol Leverandørnr. MM0037.01
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7,7',8,8'-Tetradehydro-4,5alpha:4',5'alpha-diepoxy-17,17'-dimethyl-2,2'-bimorphinanyl-3,3',6alpha,6'alpha-tetrol (Pseudomorphine; 2,2'-Bismorphine)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pseudomorphine
CAS 125-24-6
Analyt- eller komponentnavne Pseudomorphine
Forsendelsestilstand Room Temperature
Molekylær formel C34 H36 N2 O6
Certificeringer/compliance ISO 17025
Synonym Pseudomorphine (6CI,7CI,8CI), 2,2'-Dehydrodimorphine, Dehydromorphine, Oxydimorphine, Pseudomorphine (C34 alkaloid), 7,7',8,8'-Tetradehydro-4,5alpha:4',5'alpha-diepoxy-17,17'-dimethyl-2,2'-bimorphinanyl-3,3',6alpha,6'alpha-tetrol, Morphine Sulphate Imp. B (EP), Morphine Hydrochloride Imp. B (EP), Pseudomorphine, 2,2'-Bismorphine, [2,2'-Bimorphinan]-3,3',6,6'-tetrol, 7,7',8,8'-tetradehydro-4,5:4',5'-diepoxy-17,17'-dimethyl-, (5alpha,6alpha)-(5'alpha,6'alpha)-, (5alpha,6alpha)-(5'alpha,6'alpha)-7,7',8,8'-Tetradehydro-4,5:4',5'-diepoxy-17,17'-dimethyl[2,2'-bimorphinan]-3,3',6,6'-tetrol, Morphine Related Compound B, Morphine Hydrochloride Impurity B, Morphine Sulfate Impurity B, Morphine Impurity B, Morphine Impurity B
InChI formel InChI=1S/C34H36N2O6/c1-35-9-7-33-19-3-5-23(37)31(33)41-29-25(33)15(13-21(19)35)11-17(27(29)39)18-12-16-14-22-20-4-6-24(38)32-34(20,8-10-36(22)2)26(16)30(42-32)28(18)40/h3-6,11-12,19-24,31-32,37-40H,7-10,13-14H2,1-2H3/t19-,20-,21+,22+,23-,24-,31-,32-,33-,34-/m0/s1
SMIL CN1CC[C@]23[C@H]4Oc5c(O)c(cc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35)c6cc7C[C@@H]8[C@@H]9C=C[C@H](O)[C@@H]%10Oc(c6O)c7[C@]9%10CCN8C
IUPAC navn (4R,4aR,7S,7aR,12bS)-10-[(4R,4aR,7S,7aR,12bS)-7,9-dihydroxy-3-methyl-2,4,4a,7,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-10-yl]-3-methyl-2,4,4a,7,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7,9-diol
Molekylvægt (g/mol) 568.66
Mængde 100 mg
Formel vægt 568.2573 g/mol