Få mere at vide
7-(4-Bromobutoxy-1,1,2,2,3,3,4,4-d8)-2(1H)-quinolinone, TRC
Artikelnummer. 30346419
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30346419 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30346419 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68198225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-(4-Bromobutoxy-1,1,2,2,3,3,4,4-d8)-2(1H)-quinolinone
CAS 1427053-44-8
Molekylær formel C13H6D8BrNO2
Mængde 25 mg
Synonym 7-(4-Bromobutoxy)-quinoline-2(1H)-one-d8
InChI formel InChI=1S/C13H14BrNO2/c14-7-1-2-8-17-11-5-3-10-4-6-13(16)15-12(10)9-11/h3-6,9H,1-2,7-8H2,(H,15,16)/i1D2,2D2,7D2,8D2
SMIL [2H]C([2H])(Br)C([2H])([2H])C([2H])([2H])C([2H])([2H])Oc1ccc2C=CC(=O)Nc2c1
IUPAC navn 7-(4-bromo-1,1,2,2,3,3,4,4-octadeuteriobutoxy)-1H-quinolin-2-one
Molekylvægt (g/mol) 304.21
Formel vægt 303.071