Få mere at vide
7-[4-[4-(3-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one, Mikromol™
Artikelnummer. 30458475
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458475 4 x 25 mg
17940746 25 mg
17981076 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458475 Leverandør Mikromol Leverandørnr. MM0961.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-[4-[4-(3-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Deschloro Aripiprazole
CAS 203395-82-8
Analyt- eller komponentnavne 2-Deschloro Aripiprazole
Forsendelsestilstand Room Temperature
Molekylær formel C23 H28 Cl N3 O2
Certificeringer/compliance ISO 17025
Synonym Didanosine Imp. G (EP), Didanosine USP Related Compound B, Didanosine USP RC B, 2(1H)-Quinolinone, 7-[4-[4-(3-chlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-, 2-Deschloro Aripiprazole, Aripiprazol Imp. D (EP), Aripiprazole Impurity D
InChI formel InChI=1S/C23H28ClN3O2/c24-19-4-3-5-20(16-19)27-13-11-26(12-14-27)10-1-2-15-29-21-8-6-18-7-9-23(28)25-22(18)17-21/h3-6,8,16-17H,1-2,7,9-15H2,(H,25,28)
Løsningstype Neat
SMIL Clc1cccc(c1)N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2
IUPAC navn 7-[4-[4-(3-chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
Molekylvægt (g/mol) 413.94
Mængde 4 x 25 mg
Formel vægt 413.187 g/mol