missing translation for 'onlineSavingsMsg'
Få mere at vide

7-[4-[4-(3-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one, Mikromol™

Artikelnummer. 17940746
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17940746 25 mg
17981076 100 mg
30458475 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17940746 Leverandør Mikromol Leverandørnr. MM0961.070025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-[4-[4-(3-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Deschloro Aripiprazole
CAS 203395-82-8
Analyt- eller komponentnavne 2-Deschloro Aripiprazole
Forsendelsestilstand Room Temperature
Molekylær formel C23 H28 Cl N3 O2
Certificeringer/compliance ISO 17025
Synonym Didanosine Imp. G (EP), Didanosine USP Related Compound B, Didanosine USP RC B, 2(1H)-Quinolinone, 7-[4-[4-(3-chlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-, 2-Deschloro Aripiprazole, Aripiprazol Imp. D (EP), Aripiprazole Impurity D
InChI formel InChI=1S/C23H28ClN3O2/c24-19-4-3-5-20(16-19)27-13-11-26(12-14-27)10-1-2-15-29-21-8-6-18-7-9-23(28)25-22(18)17-21/h3-6,8,16-17H,1-2,7,9-15H2,(H,25,28)
SMIL Clc1cccc(c1)N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2
IUPAC navn 7-[4-[4-(3-chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
Molekylvægt (g/mol) 413.94
Mængde 25 mg
Formel vægt 413.187 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.