Få mere at vide
7-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy]quinolin-2(1H)-one, Mikromol™
Artikelnummer. 17963546
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17963546 25 mg
1 options
Artikelnummer. 17963546 Leverandør Mikromol Leverandørnr. MM0961.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy]quinolin-2(1H)-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dehydro Aripiprazole
CAS 129722-25-4
Analyt- eller komponentnavne Dehydro Aripiprazole
Forsendelsestilstand Room Temperature
Molekylær formel C23 H25 Cl2 N3 O2
Certificeringer/compliance ISO 17025
Synonym 7-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy]quinolin-2(1H)-one, Dehydroaripiprazole, Aripiprazole Related Compound G, Aripiprazole Imp E (EP), Aripiprazole Imp. E (EP), Aripiprazole USP Related Compound G, Aripiprazole USP RC G, Aripiprazole Related Compound G, Aripiprazole Impurity E
InChI formel InChI=1S/C23H25Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-9,16H,1-2,10-15H2,(H,26,29)
SMIL Clc1cccc(N2CCN(CCCCOc3ccc4C=CC(=O)Nc4c3)CC2)c1Cl
IUPAC navn 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1H-quinolin-2-one
Molekylvægt (g/mol) 446.37
Mængde 25 mg
Formel vægt 445.1324 g/mol