Learn More
7-(2-Carbomethoxyethyl)-5-(2-chlorophenyl)-thieno-1,4-diazepin-2-one, TRC
Tekniske data
Tekniske data
| Chemical Name or Material | 7-(2-Carbomethoxyethyl)-5-(2-chlorophenyl)-thieno-1,4-diazepin-2-one |
| CAS | 100827-80-3 |
| Molecular Formula | C17 H15 Cl N2 O3 S |
| Quantity | 2 mg |
| InChI Formula | InChI=1S/C17H15ClN2O3S/c1-23-15(22)7-6-10-8-12-16(11-4-2-3-5-13(11)18)19-9-14(21)20-17(12)24-10/h2-5,8H,6-7,9H2,1H3,(H,20,21) |
| SMILES | COC(=O)CCc1cc2C(=NCC(=O)Nc2s1)c3ccccc3Cl |
| IUPAC Name | methyl 3-[5-(2-chlorophenyl)-2-oxo-1,3-dihydrothieno[2,3-e][1,4]diazepin-7-yl]propanoate |
| Molecular Weight (g/mol) | 362.83 |
| Formula Weight | 362.0492 |