Få mere at vide
7-(1-Methyl-2-hydroxyethyl)guanine-d7 (Major), TRC
Artikelnummer. 30483115
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30483115 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483115 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31263210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-(1-Methyl-2-hydroxyethyl)guanine-d7 (Major)
CAS 1346604-24-7
Molekylær formel C8H4D7N5O2
Mængde 10 mg
Synonym 2-Amino-1,7-dihydro-7-(1-methyl-2-hydroxyethyl)-6H-purin-6-one-d7, N7-(1-Methyl-2-hydroxyethyl)guanine-d7
InChI formel InChI=1S/C8H11N5O2/c1-4(2-14)13-3-10-6-5(13)7(15)12-8(9)11-6/h3-4,14H,2H2,1H3,(H3,9,11,12,15)/i1D3,2D2,3D,4D
SMIL [2H]c1nc2NC(=NC(=O)c2n1C([2H])(C([2H])([2H])[2H])C([2H])([2H])O)N
IUPAC navn 2-amino-8-deuterio-7-(1,1,1,2,3,3-hexadeuterio-3-hydroxypropan-2-yl)-3H-purin-6-one
Molekylvægt (g/mol) 216.25
Formel vægt 216.1352