Få mere at vide
Viking Optical™ 6X Linen Tester Magnifier
Artikelnummer. 12836335
Encompass_Preferred
Skift visning
Oversigt:
25,4 x 3,2 mm
Skala: 25 x 1 mm
Pakningsstørrelse:
Hver
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Oversigt unitSize
12836335 Skala: 25 x 1 mm Hver
12846335 25,4 x 3,2 mm Hver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 12836335 Leverandør Viking Optical™ Leverandørnr. 5055
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Two element glass lens in a black finished metal folding mount. 6x magnification.
Tekniske data
Oversigt Skala: 25 x 1 mm
Forstørrelse 6x