Få mere at vide
(6S)-6-[2-(1,3-Benzodioxol-5-yl)ethyl]-5,6-dihydro-4-methoxy-2H-pyran-2-one, TRC
Artikelnummer. 30344726
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30344726 2.5 mg
30338415 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30344726 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2035302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (6S)-6-[2-(1,3-Benzodioxol-5-yl)ethyl]-5,6-dihydro-4-methoxy-2H-pyran-2-one
CAS 19902-91-1
Molekylær formel C15H16O5
Mængde 2.5 mg
Synonym (S)-5,6-Dihydro-4-methoxy-6-[3,4-(methylenedioxy)phenethyl]-2H-pyran-2-one, (S)-6-[2-(1,3-Benzodioxol-5-yl)ethyl]-5,6-dihydro-4-methoxy-2H-pyran-2-one, Methysticin, dihydro- (6CI), (+)-Dihydromethysticin, 7,8-Dihydromethysticin, DHM, Dihydromethysticin, Methysticin, 7,8-dihydro-, NSC 112159
InChI formel InChI=1S/C15H16O5/c1-17-12-7-11(20-15(16)8-12)4-2-10-3-5-13-14(6-10)19-9-18-13/h3,5-6,8,11H,2,4,7,9H2,1H3/t11-/m0/s1
SMIL COC1=CC(=O)O[C@@H](CCc2ccc3OCOc3c2)C1
IUPAC navn (2S)-2-[2-(1,3-benzodioxol-5-yl)ethyl]-4-methoxy-2,3-dihydropyran-6-one
Molekylvægt (g/mol) 276.28
Formel vægt 276.0998