Learn More
(6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate, 98%, BLD Pharm™
Product Code. 30946789
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30946789 25 g 25g
30933510 5 g 5g
30934964 1 g 1g
30946511 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30946789 Supplier BLD Pharm Supplier No. BD12037725g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 23325-78-2
Molecular Formula C16H19N3O5S
MDL Number MFCD00167148
Quantity 25 g
SMILES O=C(C(N12)=C(C)CS[C@]2([H])[C@H](NC([C@H](N)C3=CC=CC=C3)=O)C1=O)O.O
Molecular Weight (g/mol) 365.40
Percent Purity 98%
Chemical Name or Material (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate