Få mere at vide
(6R)-6-Ethyl-1,4-bis[(1S)-1-(4-methoxyphenyl)ethyl]-2,5-piperazinedione-3,3,6-d3, TRC
Artikelnummer. 30370703
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30370703 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370703 Leverandør Toronto Research Chemicals Leverandørnr. TRCE90008710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (6R)-6-Ethyl-1,4-bis[(1S)-1-(4-methoxyphenyl)ethyl]-2,5-piperazinedione-3,3,6-d3
CAS 1246496-66-1
Molekylær formel C24H27D3N2O4
Mængde 10 mg
InChI formel InChI=1S/C24H30N2O4/c1-6-22-24(28)25(16(2)18-7-11-20(29-4)12-8-18)15-23(27)26(22)17(3)19-9-13-21(30-5)14-10-19/h7-14,16-17,22H,6,15H2,1-5H3/t16-,17-,22+/m0/s1/i15D2,22D
SMIL [C@@H](C)(N1[C@](CC)(C(=O)N([C@@H](C)C2=CC=C(OC)C=C2)C(C1=O)([2H])[2H])[2H])C3=CC=C(OC)C=C3
IUPAC navn (R)-3-ethyl-1,4-bis((S)-1-(4-methoxyphenyl)ethyl)piperazine-2,5-dione-3,6,6-d3
Molekylvægt (g/mol) 413.52
Formel vægt 413.24