Få mere at vide
(6E)-8-Methyl-6-nonenoic Acid, TRC
Artikelnummer. 30482663
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482663 100 mg
30464352 500 mg
30473075 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482663 Leverandør Toronto Research Chemicals Leverandørnr. TRCM325790100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59320-77-3
Molekylær formel C10 H18 O2
Molekylvægt (g/mol) 170.25
IUPAC navn (E)-8-methylnon-6-enoic acid
SMIL CC(C)\C=C\CCCCC(=O)O
Tekniske data
Kemisk navn eller materiale (6E)-8-Methyl-6-nonenoic Acid
Mængde 100 mg
InChI formel InChI=1S/C10H18O2/c1-9(2)7-5-3-4-6-8-10(11)12/h5,7,9H,3-4,6,8H2,1-2H3,(H,11,12)/b7-5+
Formel vægt 170.1307