missing translation for 'onlineSavingsMsg'
Få mere at vide

6beta-Hydroxy Cortisol, TRC

Artikelnummer. 18084020
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18084020 25 mg
18053830 1 mg
18081330 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 18084020 Leverandør Toronto Research Chemicals Leverandørnr. TRCH92230025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-35-0
Molekylær formel C21 H30 O6
Molekylvægt (g/mol) 378.459
Synonym 6beta,11beta,17,21-Tetrahydroxypregn-4-ene-3,20-dione, Hydrocortisone Imp. D (EP), 6β-Hydroxyhydrocortisone, Pregn-4-ene-3,20-dione, 6,11,17,21-tetrahydroxy-, (6β,11β)-, Corticosterone, 6β,17-dihydroxy- (6CI), Pregn-4-ene-3,20-dione, 6β,11β,17,21-tetrahydroxy- (8CI), (6β,11β)-6,11,17,21-Tetrahydroxypregn-4-ene-3,20-dione, 6β,11β,17α,21-Tetrahydroxy-4-pregen-3,20-dione, 6β,11β,17α,21-Tetrahydroxypregn-4-en-3,20-dione, 6β,11β,17α,21-Tetrahydroxypregn-4-ene-3,20-dione, 6β,17-Dihydroxycorticosterone, 6β-Hydroxycortisol, NSC 76163, Hydrocortisone-6-beta-hydroxy
IUPAC navn (6R,8S,9S,10R,11S,13S,14S,17R)-6,11,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMIL C[C@]12CCC(=O)C=C1[C@H](O)C[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23

Specifications

Kemisk navn eller materiale 6beta-Hydroxy Cortisol
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C21H30O6/c1-19-5-3-11(23)7-14(19)15(24)8-12-13-4-6-21(27,17(26)10-22)20(13,2)9-16(25)18(12)19/h7,12-13,15-16,18,22,24-25,27H,3-6,8-10H2,1-2H3/t12-,13-,15+,16-,18+,19-,20-,21-/m0/s1
Formel vægt 378.204
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.