Få mere at vide
6beta-Hydroxy Budesonide, TRC
Artikelnummer. 17943395
Skift visning
Mængde:
0.25 mg
2.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17943395 0.25 mg
17942365 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17943395 Leverandør Toronto Research Chemicals Leverandørnr. TRCH8303500.25MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 88411-77-2
Molekylær formel C25 H34 O7
Molekylvægt (g/mol) 446.53
Synonym Pregna-1,4-diene-3,20-dione, 16,17-[butylidenebis(oxy)]-6,11,21-trihydroxy-, (6β,11β,16α)-, (6β,11β,16α)-16,17-[Butylidenebis(oxy)]-6,11,21-trihydroxypregna-1,4-diene-3,20-dione, 6β-Hydroxybudesonide, 6-beta-Hydroxybudesonide
SMIL CCCC1O[C@@H]2C[C@H]3[C@@H]4C[C@@H](O)C5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Tekniske data
Kemisk navn eller materiale 6Beta-Hydroxy Budesonide
Renhedsgrad noter HPLC
Mængde 0.25 mg
InChI formel InChI=1S/C25H34O7/c1-4-5-21-31-20-10-15-14-9-17(28)16-8-13(27)6-7-23(16,2)22(14)18(29)11-24(15,3)25(20,32-21)19(30)12-26/h6-8,14-15,17-18,20-22,26,28-29H,4-5,9-12H2,1-3H3/t14-,15-,17+,18-,20+,21?,22+,23-,24-,25+/m0/s1
Formel vægt 446.2305
Procent renhed >95%