Få mere at vide
6Beta-Hydroxy-21-oic Triamcinolone Acetonide, TRC
Artikelnummer. 30405915
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405915 10 mg
30407201 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30405915 Leverandør Toronto Research Chemicals Leverandørnr. TRCH97004510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68263-02-5
Molekylær formel C24 H29 F O8
Molekylvægt (g/mol) 464.48
Synonym (6β,11β,16α)-9-Fluoro-6,11-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxopregna-1,4-dien-21-oic Acid, 6β-Hydroxy-21-oic Triamcinolone Acetonide
SMIL CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@@H](O)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)C(=O)O
Tekniske data
Kemisk navn eller materiale 6Beta-Hydroxy-21-oic Triamcinolone Acetonide
Mængde 10 mg
InChI formel InChI=1S/C24H29FO8/c1-20(2)32-17-9-12-13-8-15(27)14-7-11(26)5-6-21(14,3)23(13,25)16(28)10-22(12,4)24(17,33-20)18(29)19(30)31/h5-7,12-13,15-17,27-28H,8-10H2,1-4H3,(H,30,31)/t12-,13-,15+,16-,17+,21-,22-,23-,24-/m0/s1
Formel vægt 464.1846