Få mere at vide
6Beta-Hydroxy-21-desacetyl Deflazacort, TRC
Artikelnummer. 30010622
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010622 10 mg
30006513 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010622 Leverandør Toronto Research Chemicals Leverandørnr. TRCH93580010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72099-45-7
Molekylær formel C23 H29 N O6
Molekylvægt (g/mol) 415.48
Synonym 5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 6,11,21-trihydroxy-2'-methyl-, (6β,11β,16β)- (9CI), 2H-Naphth[2',1':4,5]indeno[1,2-d]oxazole, 5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione deriv., (6β,11β,16β)-6,11,21-Trihydroxy-2'-methyl-5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 6β-Hydroxy-21-desacetyl Deflazacort, 6β-Hydroxy-21-desacetyldeflazacort
SMIL CC1=N[C@@]2([C@@H](C[C@H]3[C@@H]4C[C@@H](O)C5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C)O1)C(=O)CO
Tekniske data
Kemisk navn eller materiale 6beta-Hydroxy-21-desacetyl Deflazacort
Mængde 10 mg
InChI formel InChI=1S/C23H29NO6/c1-11-24-23(18(29)10-25)19(30-11)8-14-13-7-16(27)15-6-12(26)4-5-21(15,2)20(13)17(28)9-22(14,23)3/h4-6,13-14,16-17,19-20,25,27-28H,7-10H2,1-3H3/t13-,14-,16+,17-,19+,20+,21-,22-,23+/m0/s1
Formel vægt 415.1995