Få mere at vide
6alpha-Methylprednisolone 21-Hemisuccinate Sodium Salt, TRC
Artikelnummer. 30478911
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478911 100 mg
30472484 250 mg
30476551 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30478911 Leverandør Toronto Research Chemicals Leverandørnr. TRCM326725100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2375-03-3
Molekylær formel C26 H33 O8 . Na
Molekylvægt (g/mol) 496.53
Synonym Methylprednisolone Hydrogen Succinate Sodium Salt
IUPAC navn sodium;4-[2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoate
SMIL [Na+].C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)CCC(=O)[O-])[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
Tekniske data
Kemisk navn eller materiale Methylprednisolone Hydrogen Succinate Sodium Salt
Mængde 100 mg
InChI formel InChI=1S/C26H34O8.Na/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24;/h6,8,11,14,16-17,19,23,28,33H,4-5,7,9-10,12-13H2,1-3H3,(H,30,31);/q;+1/p-1/t14-,16-,17-,19-,23+,24-,25-,26-;/m0./s1
Formel vægt 496.2073