Få mere at vide
6Alpha-Methyl Prednisolone 17-Hemisuccinate, TRC
Artikelnummer. 30005113
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005113 10 mg
30011363 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005113 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32630010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 77074-42-1
Molekylær formel C26H34O8
Molekylvægt (g/mol) 474.54
Synonym (6α,11β)-17-(3-Carboxy-1-oxopropoxy)-11,21-dihydroxy-6-methylpregna- 1,4-diene-3,20-dione;
SMIL [H][C@]1([C@@]23[H])[C@]([C@](C=C4)(C)C([C@@H](C)C1)=CC4=O)([H])[C@@H](O)C[C@@]2([C@](C(CO)=O)(OC(CCC(O)=O)=O)CC3)C
Tekniske data
Kemisk navn eller materiale 6alpha-Methyl Prednisolone 17-Hemisuccinate
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C26H34O8/c1-14-10-16-17-7-9-26(20(30)13-27,34-22(33)5-4-21(31)32)25(17,3)12-19(29)23(16)24(2)8-6-15(28)11-18(14)24/h6,8,11,14,16-17,19,23,27,29H,4-5,7,9-10,12-13H2,1-3H3,(H,31,32)/t14-,16-,17-,19-,23+,24-,25-,26-/m0/s1
Formel vægt 474.2254
Procent renhed >95%