missing translation for 'onlineSavingsMsg'
Få mere at vide

6Alpha-Hydroxy Budesonide, TRC

Product Code. 30008919
Change view
Click to view available options
Mængde:
0.5 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30008919 1 mg
30003808 5 mg
30008507 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008919 Supplier Toronto Research Chemicals Supplier No. TRCH8303451MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577777-51-6
Molekylær formel C26 H34 O7
Molekylvægt (g/mol) 458.54
Synonym Pregna-1,4-diene-3,20-dione, 16,17-[butylidenebis(oxy)]-6,11,21-trihydroxy-, (6α,11β,16α)-, (6α,11β,16α)-16,17-[Butylidenebis(oxy)]-6,11,21-trihydroxypregna-1,4-diene-3,20-dione, 6α-Hydroxybudesonide, 6α-Hydroxy budesonide
SMIL CCC\C=C\1/O[C@@H]2C[C@H]3[C@@H]4C[C@H](O)C5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Kemisk navn eller materiale 6a-Hydroxybudesonide
Mængde 1 mg
InChI formel InChI=1S/C26H34O7/c1-4-5-6-22-32-21-11-16-15-10-18(29)17-9-14(28)7-8-24(17,2)23(15)19(30)12-25(16,3)26(21,33-22)20(31)13-27/h6-9,15-16,18-19,21,23,27,29-30H,4-5,10-13H2,1-3H3/b22-6+/t15-,16-,18-,19-,21+,23+,24-,25-,26+/m0/s1
Formel vægt 458.2305
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.