Få mere at vide
6Alpha-Chlorobeclomethasone Dipropionate, TRC
Artikelnummer. 30004759
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30004759 2.5 mg 2.5mg
30004743 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004759 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3644102.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 887130-68-9
Molekylær formel C28H36Cl2O7
Molekylvægt (g/mol) 555.49
Synonym Pregna-1,4-diene-3,20-dione, 6,9-dichloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)-, (6α,11β,16β)- (9CI), 6α-Chlorobeclomethasone dipropionate, 6α,9-Dichloro-11β-hydroxy-16β-methyl-3,20-dioxopregna-1,4-diene-17,21-diyl dipropanoate, (6α,11β,16β)-6,9-Dichloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione
IUPAC navn [2-[(6S,8S,9R,10S,11S,13S,14S,16S,17R)-6,9-dichloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMIL CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3C[C@H](Cl)C4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale 6a-Chlorobeclomethasone Dipropionate
Mængde 2.5 mg
InChI formel InChI=1S/C28H36Cl2O7/c1-6-23(34)36-14-22(33)28(37-24(35)7-2)15(3)10-17-18-12-20(29)19-11-16(31)8-9-25(19,4)27(18,30)21(32)13-26(17,28)5/h8-9,11,15,17-18,20-21,32H,6-7,10,12-14H2,1-5H3/t15-,17-,18-,20-,21-,25-,26-,27-,28-/m0/s1
Formel vægt 554.1838