Få mere at vide
6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-diene-17beta-carboxylic Acid, Mikromol™
Artikelnummer. 17912946
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17912946 25 mg
1 options
Artikelnummer. 17912946 Leverandør Mikromol Leverandørnr. MM1244.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-diene-17beta-carboxylic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fluticasone 17beta-Carboxylic Acid Propionate
CAS 65429-42-7
Analyt- eller komponentnavne Fluticasone 17beta-Carboxylic Acid Propionate
Forsendelsestilstand Room Temperature
Molekylær formel C24 H30 F2 O6
Certificeringer/compliance ISO 9001
Synonym Androsta-1,4-diene-17-carboxylic acid, 6,9-difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)-, (6alpha,11beta,16alpha,17alpha)-, 6alpha,9-difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-diene-17beta-carboxylic acid, Fluticasone Propionate Imp. A (EP), GR 36264X, Ph Eur Fluticasone Propionate Impurity A, unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), hydroxyacid propionate (GR 36264X), dithio (GR 269949X), Fluticasone Propionate Impurity A
InChI formel InChI=1S/C24H30F2O6/c1-5-19(29)32-24(20(30)31)12(2)8-14-15-10-17(25)16-9-13(27)6-7-21(16,3)23(15,26)18(28)11-22(14,24)4/h6-7,9,12,14-15,17-18,28H,5,8,10-11H2,1-4H3,(H,30,31)/t12-,14+,15+,17+,18+,21+,22+,23+,24+/m1/s1
SMIL CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)O
IUPAC navn (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylic acid
Molekylvægt (g/mol) 452.49
Mængde 25 mg
Formel vægt 452.201 g/mol